Anti-FANCC Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-194
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FANCC
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q00597
- Antigen Synonyms:bA80I15.1|FA 3|FA3|FAC|FACC|FANCC_HUMAN|Fanconi anemia complementation group C|Fanconi anemia complementation group C protein|Fanconi anemia group C protein|Fanconi pancytopenia type 3|FLJ14675|Protein FACC
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Fanconi anemia group C protein (1-210AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: FANCC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse