Anti-DNAJA1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Catalog # 77854-243
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:DNAJA1
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P31689
- Antigen Synonyms:DJ 2|DJ2|DjA1|DnaJ (Hsp40) homolog|subfamily A|member 1|DnaJ homolog subfamily A member 1|DnaJ protein homolog 2|DNAJ2|DNJA1_HUMAN|hDJ 2|HDJ-2|HDJ2|Heat shock 40 kDa protein 4|heat shock protein DNAJ like 2|Heat shock protein J2|HSDJ|HSJ-2|HSJ2|HSPF4|Human DnaJ protein 2|NEDD7|Neural precursor cell expressed developmentally down regulated 7|OTTHUMP00000021193
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human DnaJ homolog subfamily A member 1 protein (8-106AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: DNAJA1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human