Anti-FAM129A Rabbit Polyclonal Antibody
Catalog # 77848-138
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FAM129A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q9BZQ8
- Antigen Synonyms:NIBAN1|C1orf24|NIBAN|GIG39|Protein Niban 1|Cell growth-inhibiting gene 39 protein|Protein FAM129A
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence around amino acid 912~916 (K-E-G-E-G) derived from Human Niban
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:1000, IF: 1:100 to 1:200
Type:
Antigen: FAM129A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human