Anti-TRMT6 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: TRMT6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-396
- 77844-395
- Antigen Symbol:
- TRMT6
- TRMT6
- Antigen Synonyms:
- ntibody|Adenine N|CGI 09|CGI09|GCD10|Gcd10p|MGC5029|TRM6|TRM6_HUMAN|tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6|tRNA methyltransferase 6|tRNA methyltransferase 6 homolog (S. cerevisiae)|tRNA(m1A58)-methyltransferase subunit TRM6|tRNA(m1A58)MTase subunit TRM6
- ntibody|Adenine N|CGI 09|CGI09|GCD10|Gcd10p|MGC5029|TRM6|TRM6_HUMAN|tRNA (adenine-N(1)-)-methyltransferase non-catalytic subunit TRM6|tRNA methyltransferase 6|tRNA methyltransferase 6 homolog (S. cerevisiae)|tRNA(m1A58)-methyltransferase subunit TRM6|tRNA(m1A58)MTase subunit TRM6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UJA5
- Q9UJA5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6 protein (1-300AA)
- Recombinant Human tRNA (adenine(58)-N(1))-methyltransferase non-catalytic subunit TRM6 protein (1-300AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together