Anti-DSE Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: DSE
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-071
- 77844-070
- Antigen Symbol:
- DSE
- DSE
- Antigen Synonyms:
- Chondroitin glucuronate 5 epimerase|Chondroitin-glucuronate 5-epimerase|Dermatan sulfate epimerase|Dermatan-sulfate epimerase|DS epimerase|DSE_HUMAN|DSEPI|OTTHUMP00000040406|SART 2|SART-2|Squamous cell carcinoma antigen recognized by T cells 2|Squamous cell carcinoma antigen recognized by T-cells 2
- Chondroitin glucuronate 5 epimerase|Chondroitin-glucuronate 5-epimerase|Dermatan sulfate epimerase|Dermatan-sulfate epimerase|DS epimerase|DSE_HUMAN|DSEPI|OTTHUMP00000040406|SART 2|SART-2|Squamous cell carcinoma antigen recognized by T cells 2|Squamous cell carcinoma antigen recognized by T-cells 2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UL01
- Q9UL01
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human Dermatan-sulfate epimerase protein (23-270AA)
- Recombinant Human Dermatan-sulfate epimerase protein (23-270AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together