Anti-CLOCK Rabbit Recombinant Polyclonal Antibody
Catalog # 77853-274
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CLOCK
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O15516
- Antigen Synonyms:bHLHe8|Circadian locomoter output cycles kaput protein|Circadian locomoter output cycles protein kaput|Circadian Locomotor Output Cycles Kaput|Circadium Locomotor Output Cycles Kaput|Class E basic helix-loop-helix protein 8|Clock circadian regulator|Clock homolog|Clock protein|CLOCK_HUMAN|hCLOCK|KIAA0334
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Circadian locomoter output cycles protein kaput protein (577-846AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: CLOCK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human