Anti-FDFT1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-276
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FDFT1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P37268
- Antigen Synonyms:DGPT|ERG9|Farnesyl diphosphate farnesyltransferase|Farnesyl-diphosphate farnesyltransferase|FDFT_HUMAN|FPP:FPP farnesyltransferase|SQS|Squalene synthase|Squalene synthetase|SS
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Squalene synthase protein (2-417AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: FDFT1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human