Anti-PPP1R1B Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: PPP1R1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Rat
Specifications
- Catalog No:
- 77848-897
- 77848-896
- Antigen Symbol:
- PPP1R1B
- PPP1R1B
- Antigen Synonyms:
- DARPP 32|DARPP-32|Dopamine and cAMP regulated neuronal phosphoprotein 32|Dopamine and cAMP regulated neuronal phosphoprotein|Dopamine and cAMP regulated phosphoprotein|Dopamine and cAMP regulated phosphoprotein DARPP 32|Dopamine and cAMP regulated phosphoprotein DARPP32|Dopamine- and cAMP-regulated neuronal phosphoprotein|FLJ20940|IPPD|Neuronal phosphoprotein DARPP 32|PPR1B_HUMAN|Protein phosphatase 1 regulatory (inhibitor) subunit 1B|Protein phosphatase 1 regulatory inhibitor subunit 1B|Protein phosphatase 1 regulatory subunit 1B
- DARPP 32|DARPP-32|Dopamine and cAMP regulated neuronal phosphoprotein 32|Dopamine and cAMP regulated neuronal phosphoprotein|Dopamine and cAMP regulated phosphoprotein|Dopamine and cAMP regulated phosphoprotein DARPP 32|Dopamine and cAMP regulated phosphoprotein DARPP32|Dopamine- and cAMP-regulated neuronal phosphoprotein|FLJ20940|IPPD|Neuronal phosphoprotein DARPP 32|PPR1B_HUMAN|Protein phosphatase 1 regulatory (inhibitor) subunit 1B|Protein phosphatase 1 regulatory inhibitor subunit 1B|Protein phosphatase 1 regulatory subunit 1B
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Rat
- Human, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UD71
- Q9UD71
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human PPP1R1B
- Fusion protein of Human PPP1R1B
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together