Anti-BLVRB Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77851-905
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BLVRB
- Clonality:Recombinant Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:P30043
- Antigen Synonyms:Biliverdin IX beta reductase| biliverdin reductase B (flavin reductase (NADPH))| Biliverdin reductase B| Biliverdin-IX beta-reductase| BLVRB| BLVRB_HUMAN| BVR B| BVR-B| BVRB| epididymis secretory protein Li 10| Flavin reductase (NADPH)| Flavin reductase| FLR| FR| GHBP| Green heme binding protein| Green heme-binding protein| HEL-S-10| Methemoglobin reductase| MGC117413| NADPH dependent diaphorase| NADPH flavin reductase| NADPH-dependent diaphorase| NADPH-flavin reductase| SDR43U1| short chain dehydrogenase/reductase family 43U| member 1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Flavin reductase (NADPH) protein (2-206AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: BLVRB
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human