Anti-EIF4A3 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77854-615
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF4A3
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P38919
- Antigen Synonyms:ATP-dependent RNA helicase DDX48|ATP-dependent RNA helicase eIF4A-3|DDX48|DEAD box protein 48|eIF-4A-III|eIF4A-III|EIF4A3|eIF4AIII|Eukaryotic initiation factor 4A-III|Eukaryotic initiation factor 4A-like NUK-34|Eukaryotic translation initiation factor 4A isoform 3|hNMP 265|IF4A3_HUMAN|NMP 265|NMP265|Nuclear matrix protein 265|NUK34
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Eukaryotic initiation factor 4A-III protein (6-166AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EIF4A3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human