Order Entry
United States
ContactUsLinkComponent
Anti-STUB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Anti-STUB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-STUB1 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77845-191
  • 77845-190
  • Antigen Symbol:
  • STUB1
  • STUB1
  • Antigen Synonyms:
  • Antigen NY CO 7|Antigen NY-CO-7|C terminus of Hsp70-interacting protein|Carboxy terminus of Hsp70 interacting protein|Carboxy terminus of Hsp70-interacting protein|Carboxy terminus of Hsp70p interacting protein|CHIP|CHIP_HUMAN|CLL associated antigen KW 8|CLL-associated antigen KW-8|E3 ubiquitin protein ligase CHIP|E3 ubiquitin-protein ligase CHIP|Heat shock protein A binding protein 2 (c terminal)|HSPABP2|NY CO 7|PP1131|SDCCAG7|Serologically defined colon cancer antigen 7|STIP1 homology and U Box containing protein 1|STIP1 homology and U box containing protein 1 E3 ubiquitin protein ligase|STIP1 homology and U box-containing protein 1|STUB 1|UBOX 1|UBOX1
  • Antigen NY CO 7|Antigen NY-CO-7|C terminus of Hsp70-interacting protein|Carboxy terminus of Hsp70 interacting protein|Carboxy terminus of Hsp70-interacting protein|Carboxy terminus of Hsp70p interacting protein|CHIP|CHIP_HUMAN|CLL associated antigen KW 8|CLL-associated antigen KW-8|E3 ubiquitin protein ligase CHIP|E3 ubiquitin-protein ligase CHIP|Heat shock protein A binding protein 2 (c terminal)|HSPABP2|NY CO 7|PP1131|SDCCAG7|Serologically defined colon cancer antigen 7|STIP1 homology and U Box containing protein 1|STIP1 homology and U box containing protein 1 E3 ubiquitin protein ligase|STIP1 homology and U box-containing protein 1|STUB 1|UBOX 1|UBOX1
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Biotin
  • Biotin
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UNE7
  • Q9UNE7
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human E3 ubiquitin-protein ligase CHIP protein (1-48AA)
  • Recombinant Human E3 ubiquitin-protein ligase CHIP protein (1-48AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
  • Shipped on blue ice
  • Shipped on blue ice

Specifications

Frequently Bought Together