Anti-EIF4G3 Rabbit Polyclonal Antibody (Biotinylated)
Catalog # 77854-645
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:EIF4G3
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µl
- Form:Liquid
- Gene ID:O43432
- Antigen Synonyms:eIF-4-gamma 3|eIF-4-gamma II|eIF-4G 3|eIF4G 3|EIF4G3|eIF4GII|Eukaryotic translation initiation factor 4 gamma 3|eukaryotic translation initiation factor 4 gamma|3|eukaryotic translation initiation factor-4 gamma|IF4G3_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Peptide sequence from Human Eukaryotic translation initiation factor 4 gamma 3 protein (158-176AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: EIF4G3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human