Anti-FXR1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-667
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FXR1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Form:Liquid
- Gene ID:P51114
- Antigen Synonyms:Fragile X mental retardation autosomal homolog 1|Fragile X mental retardation syndrome related protein 1|Fragile X mental retardation syndrome-related protein 1|FXR1|FXR1_HUMAN|FXR1P|hFXR1p
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Fragile X mental retardation syndrome-related protein 1 protein (469-614AA)
- Purification:>95% Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:500 to 1:5000, IHC:1:20 to 1:200, IF:1:50 to 1:200
Type:
Antigen: FXR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse