Anti-ARL11 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:25 to 1:100
Type:
Antigen: ARL11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Specifications
- Catalog No:
- 77846-835
- 77846-836
- Antigen Symbol:
- ARL11
- ARL11
- Antigen Synonyms:
- ADP ribosylation factor like 11|ADP ribosylation factor like protein 11|ADP ribosylation factor like tumor suppressor protein 1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1|ARL11_HUMAN|ARLTS1
- ADP ribosylation factor like 11|ADP ribosylation factor like protein 11|ADP ribosylation factor like tumor suppressor protein 1|ADP-ribosylation factor-like protein 11|ADP-ribosylation factor-like tumor suppressor protein 1|ARL11_HUMAN|ARLTS1
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q969Q4
- Q969Q4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ARL11
- Fusion protein of Human ARL11
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together