Anti-BACE1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77851-765
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:BACE1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P56817
- Antigen Synonyms:APP beta secretase|Asp 2|ASP2|Aspartyl protease 2|BACE 1|BACE|BACE1|BACE1_HUMAN|Beta secretase 1|Beta secretase|Beta site amyloid beta A4 precursor protein cleaving enzyme|Beta site amyloid precursor protein cleaving enzyme 1|Beta site amyloid precursor protein cleaving enzyme|Beta site APP cleaving enzyme 1|Beta site APP cleaving enzyme|Beta-secretase 1|Beta-site amyloid precursor protein cleaving enzyme 1|Beta-site APP cleaving enzyme 1|FLJ90568|HSPC104|Memapsin 2|Memapsin-2|Memapsin2|Membrane associated aspartic protease 2|Membrane-associated aspartic protease 2|Transmembrane aspartic proteinase Asp2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Beta-secretase 1 protein (46-155AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: BACE1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human