PKA Inhibitor 5-24
Catalog # 80050-084
Supplier: MilliporeSigma
CAS Number:
Specifications
- Pack type:Plastic ampoule
- CAS No.:99534-03-9
- Size:500 μg
- Storage Conditions:Desiccated (hygroscopic), safe to freeze
- Protein Synonyms:Protein Kinase A Inhibitor 5-24|H-TTYADFIASGRTGRRNAIHD|PKA Inhibitor IV
- Protein/Peptide Name:PKA Inhibitor 5-24
- Purity:≥95% (HPLC)