Anti-CARS Rabbit Recombinant Polyclonal Antibody
Catalog # 77852-370
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CARS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:P49589
- Antigen Synonyms:Cysteine tRNA ligase| Cars| CysRS| Cysteine--tRNA ligase| Cysteinyl tRNA synthetase, cytoplasmic| Cysteinyl-tRNA synthetase| cytoplasmic| MGC:11246| SYCC_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Cysteine--tRNA ligase, cytoplasmic protein (201-500AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CARS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse