Anti-SLC4A7 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:30 to 1:150
Type:
Antigen: SLC4A7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77849-785
- 77849-784
- Antigen Symbol:
- SLC4A7
- SLC4A7
- Antigen Synonyms:
- Bicarbonate transporter|BT|Electroneutral Na/HCO(3) cotransporter|NBC2|NBC3|S4A7|S4A7_HUMAN|SBC2|SLC4A6|Sodium bicarbonate cotransporter 2|Sodium bicarbonate cotransporter 2b|Sodium bicarbonate cotransporter 3|Solute carrier family 4 member 7|Solute carrier family 4 sodium bicarbonate cotransporter member 7
- Bicarbonate transporter|BT|Electroneutral Na/HCO(3) cotransporter|NBC2|NBC3|S4A7|S4A7_HUMAN|SBC2|SLC4A6|Sodium bicarbonate cotransporter 2|Sodium bicarbonate cotransporter 2b|Sodium bicarbonate cotransporter 3|Solute carrier family 4 member 7|Solute carrier family 4 sodium bicarbonate cotransporter member 7
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y6M7
- Q9Y6M7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC4A7
- Synthetic peptide of Human SLC4A7
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together