Anti-CLEC4E Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: AFG Bioscience
New Product
Certificates
About this item
Type:
Antigen: CLEC4E
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-300
- 77844-301
- Antigen Symbol:
- CLEC4E
- CLEC4E
- Antigen Synonyms:
- C type lectin domain family 4 member E|C type lectin superfamily member 9|C-type (calcium dependent carbohydrate recognition domain) lectin superfamily member 9|C-type lectin domain family 4 member E|C-type lectin superfamily member 9|CLC4E_HUMAN|CLEC 4E|Clec4e|CLECSF9|Macrophage inducible C type lectin|Macrophage-inducible C-type lectin|MINCLE
- C type lectin domain family 4 member E|C type lectin superfamily member 9|C-type (calcium dependent carbohydrate recognition domain) lectin superfamily member 9|C-type lectin domain family 4 member E|C-type lectin superfamily member 9|CLC4E_HUMAN|CLEC 4E|Clec4e|CLECSF9|Macrophage inducible C type lectin|Macrophage-inducible C-type lectin|MINCLE
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULY5
- Q9ULY5
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human C-type lectin domain family 4 member E protein (41-219AA)
- Recombinant Human C-type lectin domain family 4 member E protein (41-219AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together