Anti-ADSS Rabbit Recombinant Polyclonal Antibody
77850-850
New Product
- Antigen Symbol:ADSS
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P30520
- Antigen Synonyms:ADSS2|ADSS|Adenylosuccinate synthetase isozyme 2|AMPSase 2|AdSS 2|EC 6.3.4.4|Adenylosuccinate synthetase|acidic isozyme|liver isozyme|L-type adenylosuccinate synthetase|IMP--aspartate ligase 2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Adenylosuccinate synthetase isozyme 2 protein (1-280AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:2000, IHC: 1:20 to 1:200
Type:
Antigen: ADSS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse