Order Entry
United States
ContactUsLinkComponent
Anti-MTRR Rabbit Recombinant polyclonal Antibody (Biotinylated)
Anti-MTRR Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product
Certificates
undefined
Anti-MTRR Rabbit Recombinant polyclonal Antibody (Biotinylated)
Supplier:  AFG Bioscience
New Product

Specifications

  • Catalog No:
  • 77843-751
  • 77843-750
  • Antigen Symbol:
  • MTRR
  • MTRR
  • Antigen Synonyms:
  • Methionine synthase reductase|MSR|EC 1.16.1.8
  • Methionine synthase reductase|MSR|EC 1.16.1.8
  • Clonality:
  • Recombinant Polyclonal
  • Recombinant Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rabbit
  • Rabbit
  • Gene ID:
  • Q9UBK8
  • Q9UBK8
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Human Methionine synthase reductase protein (1-725AA)
  • Recombinant Human Methionine synthase reductase protein (1-725AA)
  • Form:
  • Liquid
  • Liquid
  • Purification:
  • >95%, Protein G purified
  • >95%, Protein G purified
  • Storage Buffer:
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
  • Storage Temperature:
  • Upon receipt, store at −20 or −80°C, avoid repeated freeze
  • Upon receipt, store at −20 or −80°C, avoid repeated freeze
  • Shipping Temperature:
  • Shipped on Blue ice
  • Shipped on Blue ice

Specifications

Frequently Bought Together