Anti-BMPR1B Rabbit Recombinant Polyclonal Antibody
77851-919
New Product
- Antigen Symbol:BMPR1B
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:O00238
- Antigen Synonyms:Activin receptor like kinase 6|Acvrlk6|ALK 6|ALK6|alk6tr|BMP type-1B receptor|BMPR IB|BMPR-1B|BMPRIB|BMR1B_HUMAN|Bone morphogenetic protein receptor type 1B|Bone morphogenetic protein receptor type IB|Bone morphogenetic protein receptor type-1B|BR 1b|BR1b|CDw 293|CDw293|CDw293 antigen|CFK 43a|CFK43a|Serine/threonine receptor kinase|zALK 6|zALK6
- Storage Buffer:0.03% proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Bone morphogenetic protein receptor type-1B protein (14-126AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200, IF: 1:50 to 1:200
Type:
Antigen: BMPR1B
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat