Anti-BEX3 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200
Type:
Antigen: BEX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77847-595
- 77847-594
- Antigen Symbol:
- BEX3
- BEX3
- Antigen Synonyms:
- Bex| BEX3_HUMAN| Brain expressed X linked 3 (mouse) homolog| brain expressed| X-linked 3| Brain-expressed X-linked gene 3| Brain-expressed X-linked protein 3| DXS6984E| HGR74| NADE| Nerve growth factor receptor (TNFRSF16) associated protein 1| Nerve growth factor receptor associated protein 1| Nerve growth factor receptor-associated protein 1| NGFR-associated protein 1| NGFRAP1| Ovarian granulosa cell 13.0 kDa protein HGR74| ovarian granulosa cell protein (13kD)| Ovarian granulosa cell protein| p75NTR associated cell death executor| p75NTR-associated cell death executor| Protein BEX3
- Bex| BEX3_HUMAN| Brain expressed X linked 3 (mouse) homolog| brain expressed| X-linked 3| Brain-expressed X-linked gene 3| Brain-expressed X-linked protein 3| DXS6984E| HGR74| NADE| Nerve growth factor receptor (TNFRSF16) associated protein 1| Nerve growth factor receptor associated protein 1| Nerve growth factor receptor-associated protein 1| NGFR-associated protein 1| NGFRAP1| Ovarian granulosa cell 13.0 kDa protein HGR74| ovarian granulosa cell protein (13kD)| Ovarian granulosa cell protein| p75NTR associated cell death executor| p75NTR-associated cell death executor| Protein BEX3
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q00994
- Q00994
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NGFRAP1
- Fusion protein of Human NGFRAP1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Frequently Bought Together