Anti-MKL2 Rabbit Recombinant Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: MKL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77844-643
- 77844-644
- Antigen Symbol:
- MKL2
- MKL2
- Antigen Synonyms:
- FLJ31823|Megakaryoblastic leukemia 2|megakaryoblastic leukemia 2 protein|MKL/myocardin-like 2|MKL/myocardin-like protein 2|Mkl2|MKL2_HUMAN|MRTF-B|Myocardin-related transcription factor B|NPD001
- FLJ31823|Megakaryoblastic leukemia 2|megakaryoblastic leukemia 2 protein|MKL/myocardin-like 2|MKL/myocardin-like protein 2|Mkl2|MKL2_HUMAN|MRTF-B|Myocardin-related transcription factor B|NPD001
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULH7
- Q9ULH7
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human MKL/myocardin-like protein 2 protein (164-313AA)
- Recombinant Human MKL/myocardin-like protein 2 protein (164-313AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95% Protein G purified
- >95% Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together