Order Entry
United States
ContactUsLinkComponent
 
Anti-CNR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-346
Supplier:  AFG Bioscience
New Product
undefined
Anti-CNR2 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Catalog # 77853-346
Supplier:  AFG Bioscience
Supplier Number:  A17728-50UG
New Product

Specifications

  • Antigen Symbol:
    RAB11FIP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    50 µg
  • Form:
    Liquid
  • Gene ID:
    Q7L804
  • Antigen Synonyms:
    KIAA0941|NRip11|RAB11 family interacting protein 2 (class I)|Rab11 family-interacting protein 2|RAB11 FIP2 long isoform|Rab11-FIP2|RAB11FIP2|RFIP2|RFIP2_HUMAN
  • Storage Buffer:
    PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
  • Storage Temperature:
    Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
  • Shipping Temperature:
    Shipped on blue ice
  • Immunogen:
    KIAA0941
  • Purification:
    Antigen affinity purified

Specifications

About this item

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human