Anti-TPMT Rabbit Recombinant Polyclonal Antibody
77853-441
New Product
- Antigen Symbol:TPMT
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Form:Liquid
- Gene ID:P51580
- Antigen Synonyms:HGNC:12014|S adenosyl L methionine thiopurine S methyltransferase|Thiopurine methyltransferase|Thiopurine S methyltransferase|Thiopurine S-methyltransferase|TPMT_HUMAN
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Thiopurine S-methyltransferase protein (4-244AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB: 1:1000 to 1:5000, IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: TPMT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human