Anti-S100A6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: S100A6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-807
- 77848-806
- Antigen Symbol:
- S100A6
- S100A6
- Antigen Synonyms:
- 2A9|5B10|CABP|CACY|Calcyclin|Growth factor inducible protein 2A9|Growth factor-inducible protein 2A9|MLN 4|MLN4|OTTHUMP00000015472|OTTHUMP00000015473|PRA|PRAGrowth factor inducible protein 2A9|Prolactin receptor associated protein|Prolactin receptor-associated protein|Protein S100 A6|Protein S100-A6|S100 A6|S100 calcium binding protein A6 (calcyclin)|S100 calcium binding protein A6|S100 calcium-binding protein A6|S10A6_HUMAN
- 2A9|5B10|CABP|CACY|Calcyclin|Growth factor inducible protein 2A9|Growth factor-inducible protein 2A9|MLN 4|MLN4|OTTHUMP00000015472|OTTHUMP00000015473|PRA|PRAGrowth factor inducible protein 2A9|Prolactin receptor associated protein|Prolactin receptor-associated protein|Protein S100 A6|Protein S100-A6|S100 A6|S100 calcium binding protein A6 (calcyclin)|S100 calcium binding protein A6|S100 calcium-binding protein A6|S10A6_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P06703
- P06703
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human S100A6
- Fusion protein of Human S100A6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together