Anti-NEU2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: NEU2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-672
- 77845-671
- Antigen Symbol:
- NEU2
- NEU2
- Antigen Synonyms:
- Cytosolic sialidase|MGC129579|N acetyl alpha neuraminidase 2|N-acetyl-alpha-neuraminidase 2|Neu2|NEUR2_HUMAN|Neuraminidase 2|OTTHUMP00000164374|SIAL2|Sialidase 2|Sialidase-2
- Cytosolic sialidase|MGC129579|N acetyl alpha neuraminidase 2|N-acetyl-alpha-neuraminidase 2|Neu2|NEUR2_HUMAN|Neuraminidase 2|OTTHUMP00000164374|SIAL2|Sialidase 2|Sialidase-2
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9Y3R4
- Q9Y3R4
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Sialidase-2 protein (171-380AA)
- Recombinant Human Sialidase-2 protein (171-380AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together