Anti-CLDN14 Rabbit Recombinant Polyclonal Antibody
77853-213
New Product
- Antigen Symbol:CLDN14
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:O95500
- Antigen Synonyms:AI851731|Claudin 14|Claudin-14|CLD14_HUMAN|CLDN 14|DFNB 29|DFNB29|Human CLDN14 gene|MGC109472|OTTHUMP00000109046|OTTHUMP00000109049|OTTMUSP00000021531|UNQ777/PRO1571
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Claudin-14 protein (140-239AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:200 to 1:1000, IHC: 1:20 to 1:200
Type:
Antigen: CLDN14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat