Human Follicle Stimulating Hormone (from Pituitary)
Supplier: MilliporeSigma
Certificates
About this item
Follicle stimulating hormone, Human pituitary, Iodination grade, Follicle stimulating hormone, Human pituitary, Iodination grade, CAS 9002-68-0, is a native pituitary protein that regulates sertoli cells by acting on G-protein-linked cell surface FSH receptors.
Follicle-stimulating hormone (FSH) belongs to the family of glycoprotein hormones. It is synthesized in the gonadotrophs present in the pituitary gland. FSH contains non-covalently associated α- and β-subunits and is a disulfide-rich heterodimer. Research area: Cell Signaling
Specifications
- CAS No.:9002-68-0
- Source:Pituitary
- Species:Human
- Protein Synonyms:Iodination Grade|Follitropin|hFSH
- Protein/Peptide Name:Follicle Stimulating Hormone
- Purity:≥90%
Specifications
Frequently Bought Together