Anti-PPIF Rabbit Polyclonal Antibody
Catalog # 77847-795
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:PPIF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse,Rat
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:P30405
- Antigen Synonyms:Cyclophilin 3|cyclophilin D|Cyclophilin F|Cyp D|CyP M|CyP-D|CyP-M|CYP3|CypD|hCyP3|mitochondrial|Mitochondrial cyclophilin|Peptidyl prolyl cis trans isomerase, mitochondral|Peptidyl-prolyl cis-trans isomerase F|Peptidyl-prolyl cis-trans isomerase F, mitochondrial|Peptidylprolyl isomerase F (cyclophilin F)|Peptidylprolyl isomerase F|PPIase|PPIase F|PPIF_HUMAN|Rotamase|Rotamase F
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human PPIF.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000, IHC: 1:50 to 1:100
Type:
Antigen: PPIF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat