Anti-MOB1B Rabbit Polyclonal Antibody
Catalog # 77849-198
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:MOB1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µl
- Form:Liquid
- Gene ID:Q7L9L4
- Antigen Synonyms:MATS 2|MATS2|MGC33910|Mob 1A|Mob 1B|MOB 4A|MOB kinase activator 1B|Mob1 homolog 1A|MOB1 Mps One Binder homolog B|MOB1 Mps one binder kinase activator like 1A|Mob1A|MOBKL 1A|MOBKL1A|MOL1A_HUMAN|Mps one binder kinase activator like 1A|Mps one binder kinase activator-like 1A|Protein Mob4A
- Storage Buffer:PBS, 150 mM NaCl, 0.02% sodium azide, 50% glycerol, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Synthesized peptide derived from internal of Human MOL1A.
- Purification:Antigen affinity purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:3000
Type:
Antigen: MOB1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse