Anti-HNF1A Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000
Type:
Antigen: HNF1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Specifications
- Catalog No:
- 77848-337
- 77848-338
- Antigen Symbol:
- HNF1A
- HNF1A
- Antigen Synonyms:
- Albumin proximal factor|Hepatic nuclear factor 1 alpha|Hepatic nuclear factor 1|Hepatic transcription factor 1 alpha|Hepatic transcription factor 1|Hepatocyte nuclear factor 1-alpha|HNF 1|HNF 1A|HNF-1-alpha|HNF-1A|hnf1a|HNF1A_HUMAN|Interferon production regulator factor|LF B1|LF B1 hepatic nuclear factor|LFB 1|LFB1|LFB1 hepatic nuclear factor|Liver specific transcription factor LF B1|Liver specific transcription factor LFB1|Liver-specific transcription factor LF-B1|Maturity onset diabetes of the young 3|MODY 3|MODY3|TCF 1|TCF-1|TCF1|Transcription factor 1|Transcription factor 1 hepatic
- Albumin proximal factor|Hepatic nuclear factor 1 alpha|Hepatic nuclear factor 1|Hepatic transcription factor 1 alpha|Hepatic transcription factor 1|Hepatocyte nuclear factor 1-alpha|HNF 1|HNF 1A|HNF-1-alpha|HNF-1A|hnf1a|HNF1A_HUMAN|Interferon production regulator factor|LF B1|LF B1 hepatic nuclear factor|LFB 1|LFB1|LFB1 hepatic nuclear factor|Liver specific transcription factor LF B1|Liver specific transcription factor LFB1|Liver-specific transcription factor LF-B1|Maturity onset diabetes of the young 3|MODY 3|MODY3|TCF 1|TCF-1|TCF1|Transcription factor 1|Transcription factor 1 hepatic
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P20823
- P20823
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human HNF1A
- Synthetic peptide of Human HNF1A
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 °C or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together