Anti-SRRM2 Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: SRRM2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77845-063
- 77845-062
- Antigen Symbol:
- SRRM2
- SRRM2
- Antigen Synonyms:
- 300 kDa nuclear matrix antigen|CWC21|KIAA0324|Ser/Arg-related nuclear matrix protein of 300 kDa|Serine/arginine repetitive matrix protein 2|Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa|Splicing coactivator subunit SRm300|SR-related nuclear matrix protein of 300 kDa|SRL300|SRM300|SRRM2|SRRM2_HUMAN|Tax-responsive enhancer element-binding protein 803|TaxREB803
- 300 kDa nuclear matrix antigen|CWC21|KIAA0324|Ser/Arg-related nuclear matrix protein of 300 kDa|Serine/arginine repetitive matrix protein 2|Serine/arginine-rich splicing factor-related nuclear matrix protein of 300 kDa|Splicing coactivator subunit SRm300|SR-related nuclear matrix protein of 300 kDa|SRL300|SRM300|SRRM2|SRRM2_HUMAN|Tax-responsive enhancer element-binding protein 803|TaxREB803
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UQ35
- Q9UQ35
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human Serine/arginine repetitive matrix protein 2 protein (248-462AA)
- Recombinant Human Serine/arginine repetitive matrix protein 2 protein (248-462AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together