Anti-ACP6 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: ACP6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-811
- 77848-812
- Antigen Symbol:
- ACP6
- ACP6
- Antigen Synonyms:
- Acid phosphatase 6| Acid phosphatase 6| lysophosphatidic| Acid phosphatase like 1| Acid phosphatase like protein 1| Acid phosphatase-like protein 1| ACPL1| LPAP| Lysophosphatidic acid phosphatase 6| Lysophosphatidic acid phosphatase type 6| Lysophosphatidic acid phosphatase type 6 precursor| PACPL1| PPA6_HUMAN
- Acid phosphatase 6| Acid phosphatase 6| lysophosphatidic| Acid phosphatase like 1| Acid phosphatase like protein 1| Acid phosphatase-like protein 1| ACPL1| LPAP| Lysophosphatidic acid phosphatase 6| Lysophosphatidic acid phosphatase type 6| Lysophosphatidic acid phosphatase type 6 precursor| PACPL1| PPA6_HUMAN
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9NPH0
- Q9NPH0
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human ACP6
- Fusion protein of Human ACP6
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Upon receipt, store at −20 or −80 °C, avoid repeated freeze
- Shipping Temperature:
- Shipped on Blue ice
- Shipped on Blue ice
Specifications
Frequently Bought Together