Anti-NME8 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA:1:2000 to 1:5000, IHC:1:25 to 1:100
Type:
Antigen: NME8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77843-377
- 77843-376
- Antigen Symbol:
- NME8
- NME8
- Antigen Synonyms:
- NME8|SPTRX2|TXNDC3|Thioredoxin domain-containing protein 3|NM23-H8|NME/NM23 family member 8|Spermatid-specific thioredoxin-2|Sptrx-2
- NME8|SPTRX2|TXNDC3|Thioredoxin domain-containing protein 3|NM23-H8|NME/NM23 family member 8|Spermatid-specific thioredoxin-2|Sptrx-2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N427
- Q8N427
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Fusion protein of Human NME8
- Fusion protein of Human NME8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together