Anti-ALAS1 Rabbit Recombinant polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77850-992
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:ALAS1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P13196
- Antigen Synonyms:5 aminolevulinate synthase |5 aminolevulinate synthase nonspecific mitochondrial|5 aminolevulinic acid synthase|5-aminolevulinate synthase|5-aminolevulinic acid synthase 1|Alas 1|ALAS 3|ALAS|ALAS H|ALAS HOUSEKEEPING TYPE|ALAS N|ALAS-H|ALAS3|ALASH|Aminolevulinate delta synthase 1|Aminolevulinic acid synthase 1|Delta ALA synthetase|Delta aminolevulinate synthase|Delta-ALA synthase 1|Delta-aminolevulinate synthase 1|HEM1_HUMAN|MIG 4|MIG4|Migration inducing protein 4|mitochondrial|nonspecific
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at −20 or −80°C, avoid repeated freeze
- Shipping Temperature:Shipped on Blue ice
- Immunogen:Recombinant Human 5-aminolevulinate synthase, nonspecific, mitochondrial protein (57-285AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
0
Type:
Antigen: ALAS1
Clonality: Recombinant polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human