Anti-CACNA2D1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # 77852-256
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:CACNA2D1
- Clonality:Recombinant Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:P54289
- Antigen Synonyms:CA2D1_HUMAN|CACN A2|CACNA2|CACNL2A|Calcium channel L type alpha 2 polypeptide|Calcium channel voltage dependent alpha 2/delta subunit 1|CCHL2A|Dihydropyridine receptor alpha 2 subunit|Dihydropyridine sensitive L type calcium channel alpha 2/delta subunit|Dihydropyridine sensitive L type calcium channel subunits alpha 2/delta|L type calcium channel subunit alpha 2|MHS 3|MHS3|Voltage dependent calcium channel subunit alpha 2/delta 1|Voltage gated calcium channel subunit alpha 2/delta 1|Voltage-dependent calcium channel subunit delta-1|Voltage-gated calcium channel subunit alpha-2/delta-1
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Voltage-dependent calcium channel subunit alpha-2/delta-1 protein (577-717AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Type:
Antigen: CACNA2D1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human