Anti-CGGBP1 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
New Product
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CGGBP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-804
- 77843-805
- Antigen Symbol:
- CGGBP1
- CGGBP1
- Antigen Synonyms:
- 20 kDa CGG binding protein| 20 kDa CGG-binding protein| CGBP1_HUMAN| CGG binding protein 1| CGG triplet repeat binding protein 1| CGG triplet repeat-binding protein 1| CGG-binding protein 1| CGGBP 1| CGGBP| Cggbp1| OTTHUMP00000213853| OTTHUMP00000213877| p20 CGG binding protein| p20 CGGBP| p20 CGGBP DNA binding protein| p20-CGGBP DNA-binding protein
- 20 kDa CGG binding protein| 20 kDa CGG-binding protein| CGBP1_HUMAN| CGG binding protein 1| CGG triplet repeat binding protein 1| CGG triplet repeat-binding protein 1| CGG-binding protein 1| CGGBP 1| CGGBP| Cggbp1| OTTHUMP00000213853| OTTHUMP00000213877| p20 CGG binding protein| p20 CGGBP| p20 CGGBP DNA binding protein| p20-CGGBP DNA-binding protein
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- HRP (Horseradish Peroxidase)
- HRP (Horseradish Peroxidase)
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UFW8
- Q9UFW8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Recombinant Human CGG triplet repeat-binding protein 1 protein (1-167AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together