Anti-SLC43A2 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: ELISA: 1:1000 to 1:2000, WB: 1:200 to 1:1000
Type:
Antigen: SLC43A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Specifications
- Catalog No:
- 77847-355
- 77847-354
- Antigen Symbol:
- SLC43A2
- SLC43A2
- Antigen Synonyms:
- FLJ23848|L-type amino acid transporter 4|Large neutral amino acids transporter small subunit 4|LAT4|LAT4_HUMAN|MGC34680|Solute carrier family 43 (amino acid system L transporter) member 2|Solute carrier family 43 member 2
- FLJ23848|L-type amino acid transporter 4|Large neutral amino acids transporter small subunit 4|LAT4|LAT4_HUMAN|MGC34680|Solute carrier family 43 (amino acid system L transporter) member 2|Solute carrier family 43 member 2
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse
- Human, Mouse
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q8N370
- Q8N370
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human SLC43A2
- Synthetic peptide of Human SLC43A2
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together