Anti-DPP6 Rabbit Recombinant Polyclonal Antibody (HRP (Horseradish Peroxidase))
77854-335
New Product
- Antigen Symbol:DPP6
- Clonality:Recombinant Polyclonal
- Conjugation:HRP (Horseradish Peroxidase)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:P42658
- Antigen Synonyms:DPP6Dipeptidyl aminopeptidase-like protein 6| DPPX| Dipeptidyl aminopeptidase-related protein| Dipeptidyl peptidase 6| Dipeptidyl peptidase IV-like protein| Dipeptidyl peptidase VI| DPP VI
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Dipeptidyl aminopeptidase-like protein 6 protein (117-859AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: DPP6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human