Anti-ALDH1B1 Rabbit Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: ALDH1B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77848-247
- 77848-246
- Antigen Symbol:
- ALDH1B1
- ALDH1B1
- Antigen Synonyms:
- Acetaldehyde dehydrogenase 5|AL1B1_HUMAN|Aldehyde dehydrogenase 1 family|member B1|Aldehyde dehydrogenase 1B1|Aldehyde dehydrogenase 5|Aldehyde dehydrogenase family 1 member B1|Aldehyde dehydrogenase X|Aldehyde dehydrogenase X mitochondrial|ALDH class 2|Aldh1b1|ALDH5|ALDHX|MGC2230|Mitochondrial aldehyde dehydrogenase X|mitochondrial|OTTHUMP00000021399
- Acetaldehyde dehydrogenase 5|AL1B1_HUMAN|Aldehyde dehydrogenase 1 family|member B1|Aldehyde dehydrogenase 1B1|Aldehyde dehydrogenase 5|Aldehyde dehydrogenase family 1 member B1|Aldehyde dehydrogenase X|Aldehyde dehydrogenase X mitochondrial|ALDH class 2|Aldh1b1|ALDH5|ALDHX|MGC2230|Mitochondrial aldehyde dehydrogenase X|mitochondrial|OTTHUMP00000021399
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- P30837
- P30837
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human ALDH1B1
- Synthetic peptide of Human ALDH1B1
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together