Anti-PCDHA6 Rabbit Recombinant Polyclonal Antibody
New Product
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: PCDHA6
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77843-956
- 77843-955
- Antigen Symbol:
- PCDHA6
- PCDHA6
- Antigen Synonyms:
- Cadherin related neuronal receptor 2|CNR 2|CNR2|CNRN 2|CNRN2|CNRS 2|CNRS2|CRNR 2|CRNR2|KIAA0345 like 8|PCDA6_HUMAN|PCDH alpha6|PCDH-alpha-6|PCDHA 6|Protocadherin alpha 6|Protocadherin alpha-6
- Cadherin related neuronal receptor 2|CNR 2|CNR2|CNRN 2|CNRN2|CNRS 2|CNRS2|CRNR 2|CRNR2|KIAA0345 like 8|PCDA6_HUMAN|PCDH alpha6|PCDH-alpha-6|PCDHA 6|Protocadherin alpha 6|Protocadherin alpha-6
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UN73
- Q9UN73
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant human Protocadherin alpha-6 protein (1-230AA)
- Recombinant human Protocadherin alpha-6 protein (1-230AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together