Anti-GRM8 Rabbit Polyclonal Antibody
New Product
Recommended dilution: ELISA: 1:1000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200
Type:
Antigen: GRM8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 77850-030
- 77850-031
- Antigen Symbol:
- GRM8
- GRM8
- Antigen Synonyms:
- FLJ41058|GLUR8|Glutamate receptor metabotropic 8|GPRC1H|GRM8_HUMAN|Metabotropic glutamate receptor 8|MGC126724|mGlu8|mGluR8
- FLJ41058|GLUR8|Glutamate receptor metabotropic 8|GPRC1H|GRM8_HUMAN|Metabotropic glutamate receptor 8|MGC126724|mGlu8|mGluR8
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human, Mouse, Rat
- Human, Mouse, Rat
- Host:
- Rabbit
- Rabbit
- Gene ID:
- O00222
- O00222
- Isotype:
- IgG
- IgG
- Western Blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide of Human GRM8
- Synthetic peptide of Human GRM8
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- PBS, 0.05% sodium azide, 40% glycerol, pH 7.4
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together