Anti-VANGL2 Rabbit Recombinant Polyclonal Antibody
77843-926
New Product
- Antigen Symbol:VANGL2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:Q9ULK5
- Antigen Synonyms:Homolog of Drosophila strabismus|KIAA1215|Loop tail associated protein|loop tail protein 1 homolog|Loop-tail protein 1 homolog|LPP 1|LPP1|LTAP|MGC119403|MGC119404|STB 1|STB1|STBM 1|STBM|STBM1|Strabismus 1|van Gogh like protein 2|Van Gogh-like protein 2|Vang (van gogh, Drosophila) like 2|Vang like 2 (van gogh, Drosophila)|vang like 2|vang like protein 2|Vang-like protein 2|VANG2_HUMAN|VANGL 2|VANGL planar cell polarity protein 2|Vangl2
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human Vang-like protein 2 protein (1-110AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:20 to 1:200
Type:
Antigen: VANGL2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse