Anti-FGL1 Rabbit Recombinant Polyclonal Antibody
Catalog # 77855-407
Supplier: AFG Bioscience
New Product
Specifications
- Antigen Symbol:FGL1
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µg
- Form:Liquid
- Gene ID:Q08830
- Antigen Synonyms:FGL 1|FGL1_HUMAN|Fibrinogen like 1|Fibrinogen like protein 1|Fibrinogen related protein 1|Fibrinogen-like protein 1|Hepassocin|Hepatocellular carcinoma related sequence|Hepatocyte derived fibrinogen related protein 1|Hepatocyte-derived fibrinogen-related protein 1|HFREP 1|HFREP-1|HFREP1|HP 041|HP-041|HP041|LFIRE 1|LFIRE-1|LFIRE1|Liver fibrinogen related protein 1|Liver fibrinogen-related protein 1|MFIRE 1|MGC108569|MGC12455|MGC37822|OTTHUMP00000122468
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Fibrinogen-like protein 1 protein (179-282AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Specifications
About this item
Recommended dilution: WB: 1:500 to 1:5000, IHC: 1:200 to 1:500
Type:
Antigen: FGL1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human