Anti-SHANK2 Rabbit Recombinant Polyclonal Antibody (Biotin)
New Product
Type:
Antigen: SHANK2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- 77845-231
- 77845-230
- Antigen Symbol:
- SHANK2
- SHANK2
- Antigen Synonyms:
- CORTBP1|KIAA1022|PROSAP1|SH3 and multiple ankyrin repeat domains protein 2|Cortactin-binding protein 1|Proline-rich synapse-associated protein 1
- CORTBP1|KIAA1022|PROSAP1|SH3 and multiple ankyrin repeat domains protein 2|Cortactin-binding protein 1|Proline-rich synapse-associated protein 1
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Biotin
- Biotin
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9UPX8
- Q9UPX8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Immunogen:
- Recombinant human SH3 and multiple ankyrin repeat domains protein 2 protein (728-822AA)
- Recombinant human SH3 and multiple ankyrin repeat domains protein 2 protein (728-822AA)
- Form:
- Liquid
- Liquid
- Purification:
- >95%, Protein G purified
- >95%, Protein G purified
- Storage Buffer:
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Frequently Bought Together