Anti-CLASP2 Rabbit Recombinant Polyclonal Antibody
77853-176
New Product
- Antigen Symbol:CLASP2
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Size:50 µg
- Form:Liquid
- Gene ID:O75122
- Antigen Synonyms:CLAP2_HUMAN|CLASP 2|CLIP associating protein 2|CLIP-associating protein 2|Cytoplasmic linker associated protein 2|Cytoplasmic linker-associated protein 2|hOrbit2|KIAA0627|Multiple asters (Mast) like homolog 2|Protein Orbit homolog 2
- Storage Buffer:0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
- Storage Temperature:Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant human CLIP-associating protein 2 protein (226-381AA)
- Purification:>95%, Protein G purified
- Packaging:Vial
Recommended dilution: IHC: 1:20 to 1:200, IP: 1:200 to 1:2000
Type:
Antigen: CLASP2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human