Anti-CBLC Rabbit Recombinant Polyclonal Antibody
Supplier: AFG Bioscience
New Product
Certificates
About this item
Recommended dilution: IHC: 1:20 to 1:200
Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- 77844-963
- 77844-964
- Antigen Symbol:
- CBLC
- CBLC
- Antigen Synonyms:
- CBLC|CBL3|RNF57E3 ubiquitin-protein ligase CBL-C|EC 2.3.2.27|RING finger protein 57|RING-type E3 ubiquitin transferase CBL-C|SH3-binding protein CBL-3|SH3-binding protein CBL-C|Signal transduction protein CBL-C
- CBLC|CBL3|RNF57E3 ubiquitin-protein ligase CBL-C|EC 2.3.2.27|RING finger protein 57|RING-type E3 ubiquitin transferase CBL-C|SH3-binding protein CBL-3|SH3-binding protein CBL-C|Signal transduction protein CBL-C
- Clonality:
- Recombinant Polyclonal
- Recombinant Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- Q9ULV8
- Q9ULV8
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant Human E3 ubiquitin-protein ligase CBL-C protein (245-474AA)
- Recombinant Human E3 ubiquitin-protein ligase CBL-C protein (245-474AA)
- Form:
- Liquid
- Liquid
- Purification:
- Antigen affinity purified
- Antigen affinity purified
- Storage Buffer:
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- PBS, 0.02% sodium azide, 50% glycerol, pH 7.3
- Storage Temperature:
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:
- Shipped on blue ice
- Shipped on blue ice
Specifications
Frequently Bought Together