Anti-CFH Rabbit Recombinant Polyclonal Antibody
77853-015
New Product
- Antigen Symbol:CFH
- Clonality:Recombinant Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:P08603
- Antigen Synonyms:adrenomedullin binding protein| age related maculopathy susceptibility 1| AHUS 1| AHUS1| AMBP 1| AMBP1| ARMD 4| ARMD4| ARMS 1| ARMS1| beta 1 H globulin| beta 1H| beta1H| CFAH_HUMAN| CFH| CFHL 3| CFHL3| Complement factor H| complement factor H| isoform b| Factor H| factor H like 1| FH| FHL 1| FHL1| H factor 1 (complement)| H factor 1| H factor 2 (complement)| HF 1| HF 2| HF| HF1| HF2| HUS| MGC88246
- Storage Buffer:PBS, 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
- Shipping Temperature:Shipped on blue ice
- Immunogen:Recombinant Human Complement factor H protein (20-270AA)
- Purification:Antigen affinity purified
- Packaging:Vial
Recommended dilution: WB:1:1000 to 1:5000
Type:
Antigen: CFH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human